Please login to your online account to display your discounted pricing

PPPA, Tocris

Competitive NR2A antagonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material PPPA
Assay Percent Range >98%
Molecular Formula C9H18NO5P
Formula Weight 251.22
Synonym (2R™, 4S™)-4-(3-Phosphonopropyl)-2-piperidinecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

227410

 
r & d systems
RDS227410
10mg Each for $257.85

227450

 
r & d systems
RDS227450
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 251.22
Quantity 10mg
Chemical Name or Material PPPA
Assay Percent Range >98%
Molecular Formula C9H18NO5P
CAS 113190-92-4
Synonym (2R™, 4S™)-4-(3-Phosphonopropyl)-2-piperidinecarboxylic acid
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature