Please login to your online account to display your discounted pricing

PPNDS, Tocris


Potent, selective P2X1 antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS130950

 View more versions of this product

Catalog No. 130950

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 694.36
Quantity 50mg
Chemical Name or Material PPNDS
Assay Percent Range >98%
Molecular Formula C18H11N4Na4O14PS2
CAS 1021868-77-8
Synonym Pyridoxal-5'-phosphate-6-(2'-naphthylazo-6'-nitro-4', 8'-disulfonate) tetrasodium salt
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material PPNDS
Assay Percent Range >98%
Molecular Formula C18H11N4Na4O14PS2
Formula Weight 694.36
Synonym Pyridoxal-5'-phosphate-6-(2'-naphthylazo-6'-nitro-4', 8'-disulfonate) tetrasodium salt
Accessories

Desiccate at -20°C