HF Scientific™ PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Use with any DPD-based photometer or test kit
Manufacturer:  HF Scientific™ HF10448A
 View more versions of this product
Catalog No. 15-393-70
Description
Description
Portable dispenser accurately delivers the correct volume of DPD reagent for Free or Total Chlorine—use with any DPD-based photometer or test kit.
  • Supplied with enough DPD reagent for 400 tests
  • To use, simply invert dispenser over sample and press button to dispense reagent; powder dissolves quickly in sample, leaving no messy residue behind
  • Available for 5 and 10mL sample sizes, with formulations for a variety of photometers and test kits
Specifications
Specifications
For total chlorine testing
400 Tests
10mL
For Hach™ and Hanna photometers
0.33 oz.
Chlorine Reagent Powder Pop Dispenser
Documents