missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HF Scientific™ PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Use with any DPD-based photometer or test kit
Supplier: HF Scientific™ HF10446A
Description
Portable dispenser accurately delivers the correct volume of DPD reagent for Free or Total Chlorine—use with any DPD-based photometer or test kit.
- Supplied with enough DPD reagent for 400 tests
- To use, simply invert dispenser over sample and press button to dispense reagent; powder dissolves quickly in sample, leaving no messy residue behind
- Available for 5 and 10mL sample sizes, with formulations for a variety of photometers and test kits
Specifications
5 mL | |
HF scientific and Hach™ photometers | |
400 Tests | |
0.16 oz. |
For chlorine testing | |
Pocket photometer | |
Chlorine Reagent Powder Pop Dispenser |