Please login to your online account to display your discounted pricing

PPADS tetrasodium salt, Tocris


P2 purinergic antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS062510

 View more versions of this product

Catalog No. 062510

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 599.3
Chemical Name or Material PPADS tetrasodium salt
Synonym Pyridoxalphosphate-6-azophenyl-2', 4'-disulfonic acid tetrasodium salt
Molecular Formula C14H10N3Na4O12PS2
Assay Percent Range >98%
CAS 192575-19-2
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material PPADS tetrasodium salt
Assay Percent Range >98%
Molecular Formula C14H10N3Na4O12PS2
Formula Weight 599.3
Synonym Pyridoxalphosphate-6-azophenyl-2', 4'-disulfonic acid tetrasodium salt
Accessories

Desiccate at -20°C