Please login to your online account to display your discounted pricing

PPADS tetrasodium salt, Tocris

P2 purinergic antagonist

$97.76 - $410.62

Chemical Identifiers

Chemical Name or Material PPADS tetrasodium salt
Assay Percent Range >98%
Molecular Formula C14H10N3Na4O12PS2
Formula Weight 599.3
Synonym Pyridoxalphosphate-6-azophenyl-2', 4'-disulfonic acid tetrasodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

062510

 
r & d systems
RDS062510
10mg Each for $97.76

062550

 
r & d systems
RDS062550
50mg Each for $410.62
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 599.3
Chemical Name or Material PPADS tetrasodium salt
Synonym Pyridoxalphosphate-6-azophenyl-2', 4'-disulfonic acid tetrasodium salt
Molecular Formula C14H10N3Na4O12PS2
Assay Percent Range >98%
CAS 192575-19-2
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C