Please login to your online account to display your discounted pricing

PP 242, Tocris™

Dual mTORC1/mTORC2 inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material PP 242
Assay Percent Range >98%
Molecular Formula C16H16N6O
Formula Weight 308.34
Synonym 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3, 4-d]pyrimidin-3-yl]-1H-indol-5-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

425710

 
r & d systems
425710
10mg Each for $290.95

425750

 
r & d systems
425750
50mg Each for $1,205.80
Description & Specifications

Specifications

Formula Weight 308.34
Quantity 10mg
CAS 1092351-67-1
Molecular Formula C16H16N6O
Chemical Name or Material PP 242
Assay Percent Range >98%
Synonym 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3, 4-d]pyrimidin-3-yl]-1H-indol-5-ol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C