Please login to your online account to display your discounted pricing

PP 121, Tocris™

Dual kinase inhibitor; inhibits PI 3-K family kinases

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material PP 121
Assay Percent Range >99%
Molecular Formula C17H17N7
Formula Weight 319.36
Synonym 1-Cyclopentyl-3-(1H-pyrrolo[2, 3-b]pyridin-5-yl)-1H-pyrazolo[3, 4-d]pyrimidin-4-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

389410

 
r & d systems
389410
10mg Each for $214.12

389450

 
r & d systems
389450
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 319.36
Quantity 10mg
Molecular Formula C17H17N7
CAS 1092788-83-4
Assay Percent Range >99%
Chemical Name or Material PP 121
Synonym 1-Cyclopentyl-3-(1H-pyrrolo[2, 3-b]pyridin-5-yl)-1H-pyrazolo[3, 4-d]pyrimidin-4-amine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C