missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Poxygrid Eye Wash Bottle Wall Rack

Holds bottle in view for emergency use.

Supplier:  Bel-Art™ H248590000

Catalog No. 10-029-254




Description

Description

Holds bottle in view for emergency use.

  • Fabricated from welded steel wire with corrosion-proof epoxy coating
  • Holds single 1000mL (32 oz.) eye wash bottle
  • For use with sterile saline eye wash and eye wash bottles (sold separately)
  • Hangs on screw or nail
Specifications

Specifications

Eye wash bottle wall rack
Epoxy Coated Steel Wire
Single 1000mL eye wash bottle
Dia. x H: 10.8 x 21 cm
SDS
Documents

Documents

Promotions

Promotions