missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ PowerUp™ SYBR™ Green & MicroAmp™ 96-well Plates Bundle DFS Item

Bundle and save up to 50% off PowerUp SYBR Green Master Mix and Plates

Manufacturer:  Applied Biosystems™ A46932

Catalog No. A46932



Description

Description

  • Reliable performance
  • Highly reproducible
  • Compatible with Applied Biosystems real-time PCR instruments
Specifications

Specifications

SYBR
Real Time PCR (qPCR)
Store at 2–8°C
DNA
PowerUp™ SYBR™ Green (2 x 5 mL) and 10 MicroAmp™ 96-well Plates
SDS
Documents
Product Certifications
Promotions

Promotions