Please login to your online account to display your discounted pricing

Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter


These electrical power cords have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.

Manufacturer: thermo scientific™  84858

 View more versions of this product

Catalog No. PI84858

  • / Each
 

Description & Specifications

Specifications

Description Power Cord with C/13 Connector, North America
Formulation Black 3m power cord
Quantity 1 cord
Sufficient For Replacement of myECL Imager and Pierce G2 Fast Blotter accessory
Item Description Power Cord with C/13 Connector, North America

Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.

Additional Information and Online Ordering »

The black, 2.4- to 3.5m-long, cords have been validated to replace the cordset that is supplied with particular Thermo Scientific powered devices. Specifically, these instruments include the myECL Imager (Part No. 62236) and the Pierce G2 Fast Blotter (Part No. 62288).

Highlights:

  • Black
  • 2.4 to 3.5m long
  • C13 connector
  • Country-specific connectors