missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Povidone-Iodine Solution USP, 10% (w/v), 1% (w/v) Available Iodine, for Laboratory Use, Ricca Chemical

$132.58 - $132.58

Chemical Identifiers

CAS 7732-18-5,25655-41-8
CAS Min % 90.0,7.93
CAS Max % 94.0,8.25
Physical Form Liquid
Chemical Name or Material Povidone-Iodine Solution, 1% (w/v) Available Iodine
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
3955-16 Ricca Chemical Company
395516
Amber Poly 500mL Each for $132.58
3955-120 Ricca Chemical Company
RI3955120
Tote 120 gal. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For Laboratory Use
Description & Specifications

Chemical Identifiers

7732-18-5,25655-41-8
94.0,8.25
Povidone-Iodine Solution, 1% (w/v) Available Iodine
90.0,7.93
Liquid
USP

Specifications

7732-18-5,25655-41-8
94.0,8.25
Amber Poly
Dark brown
Povidone-Iodine Solution, 1% (w/v) Available Iodine
90.0,7.93
Liquid
10% (w/v)
500mL
USP