missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium vinyltrifluoroborate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 387700010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Potassium vinyltrifluoroborate | |
Authentic | |
Glass bottle | |
MFCD02093335 | |
potassium vinyltrifluoroborate, potassium trifluoro vinyl borate, potassium ethenyltrifluoroboranuide, vinyltrifluoroboric acid potassium salt, potassium vinyltrifluorborate, potassium ethenyl trifluoroborate, potassium ethenyltrifluoroborate, potassium trifluoro vinyl boranuide, pubchem11308, potassiumvinyltrifluoroborate | |
[B-](C=C)(F)(F)F.[K+] | |
133.95 | |
133.95 | |
Technical |
13682-77-4 | |
94% min | |
C2H3BF3K | |
1g | |
ZCUMGICZWDOJEM-UHFFFAOYSA-N | |
potassium;ethenyl(trifluoro)boranuide | |
23679353 | |
95% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO â Research Use Only