missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium tungsten oxide, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1403114

 View more versions of this product

Catalog No. AA1403114



Specifications

Specifications

Potassium tungsten oxide
(metals basis)
MFCD00049668
potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w, potassium tungstate
AAQNGTNRWPXMPB-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)tungsten
3084032
Powder
3.12g/mL
Odorless
921°C
7790-60-5
K2O4W
14,7694
Soluble in water
[O-][W](=O)(=O)[O-].[K+].[K+]
326.033
326.04
99.5%
−60 Mesh
Hygroscopic
25g

Chemical Identifiers

7790-60-5
326.033
AAQNGTNRWPXMPB-UHFFFAOYSA-N
3084032
[O-][W](=O)(=O)[O-].[K+].[K+]
K2O4W
MFCD00049668
potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w, potassium tungstate
dipotassium;dioxido(dioxo)tungsten
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents