missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tungstate, 94%, -100 mesh, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446870250

 View more versions of this product

Catalog No. AC446870250



Description

Description

Specifications

Specifications

Potassium tungstate
53 to 56% (W)
K2WO4
2k.wo4, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate, potassium tungstate vi, potassium tungstate-w, t6vrm0ou6v, tungstate wo42-, dipotassium, t-4, unii-t6vrm0ou6v
[O-][W](=O)(=O)[O-].[K+].[K+]
326.05
326.05
Glass Bottle
25g
7790-60-5
K2O4W
15,7811
AAQNGTNRWPXMPB-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)tungsten
3084032
94%
921°C

Chemical Identifiers

7790-60-5
326.05
2k.wo4, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate, potassium tungstate vi, potassium tungstate-w, t6vrm0ou6v, tungstate wo42-, dipotassium, t-4, unii-t6vrm0ou6v
dipotassium;dioxido(dioxo)tungsten
K2O4W
AAQNGTNRWPXMPB-UHFFFAOYSA-N
3084032
[O-][W](=O)(=O)[O-].[K+].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 232-215-2

SDS
Documents