Please login to your online account to display your discounted pricing

Potassium tungstate, 94%, -100 mesh, ACROS Organics™

$119.11 - $294.88

Chemical Identifiers

Chemical Name or Material Potassium tungstate
Assay Percent Range 94%
Molecular Formula K2O4W
Formula Weight 326.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC446871000

 
thermo fisher scientific
AC446871000
100g Each for $294.88

AC446870250

 
thermo fisher scientific
AC446870250
25g Each for $119.11
Description & Specifications

Specifications

Chemical Name or Material Potassium tungstate
CAS 7790-60-5
Assay Percent Range 94%
Molecular Formula K2O4W
Formula Weight 326.04
Quantity 100g