missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium trimethylacetate, 95%, Thermo Scientific Chemicals
$80.62 - $265.79
Chemical Identifiers
CAS | 19455-23-3 |
---|---|
Molecular Formula | C5H9KO2 |
Molecular Weight (g/mol) | 140.223 |
MDL Number | MFCD00671345 |
InChI Key | WFMNHCSATCWAAQ-UHFFFAOYSA-M |
Synonym | potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt |
PubChem CID | 23662159 |
IUPAC Name | potassium;2,2-dimethylpropanoate |
SMILES | CC(C)(C)C(=O)[O-].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6280314
|
Thermo Scientific Chemicals
H6280314 |
25 g |
Each for $80.62
|
|
|||||
AAH6280322
|
Thermo Scientific Chemicals
H6280322 |
100 g |
Each for $265.79
|
|
|||||
Description
Potassium trimethylacetate is used as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium trimethylacetate is used as a intermediate for chemical research.
Notes
Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
19455-23-3 | |
140.223 | |
WFMNHCSATCWAAQ-UHFFFAOYSA-M | |
23662159 | |
CC(C)(C)C(=O)[O-].[K+] |
C5H9KO2 | |
MFCD00671345 | |
potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt | |
potassium;2,2-dimethylpropanoate |
Specifications
19455-23-3 | |
C5H9KO2 | |
MFCD00671345 | |
potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt | |
CC(C)(C)C(=O)[O-].[K+] | |
140.223 | |
140.22 | |
Potassium trimethylacetate |
∼200°C | |
(CH3)3CCO2K | |
25 g | |
WFMNHCSATCWAAQ-UHFFFAOYSA-M | |
potassium;2,2-dimethylpropanoate | |
23662159 | |
95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only