missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Trimethoxy(trifluoromethyl)borate 95.0+%, TCI America™
Supplier: TCI America P203625G
Specifications
Potassium Trimethoxy(trifluoromethyl)borate | |
118°C | |
C4H9BF3KO3 | |
25 g | |
XQUUVPNLRZQWNF-UHFFFAOYSA-N | |
potassium;trimethoxy(trifluoromethyl)boranuide | |
23682739 | |
≥95.0% (T) |
626232-27-7 | |
White | |
MFCD22124929 | |
Potassium (Trifluoromethyl)trimethoxyborate | |
[B-](C(F)(F)F)(OC)(OC)OC.[K+] | |
212.017 | |
212.02 | |
Crystalline Powder |
Chemical Identifiers
626232-27-7 | |
212.017 | |
XQUUVPNLRZQWNF-UHFFFAOYSA-N | |
23682739 | |
[B-](C(F)(F)F)(OC)(OC)OC.[K+] |
C4H9BF3KO3 | |
MFCD22124929 | |
Potassium (Trifluoromethyl)trimethoxyborate | |
potassium;trimethoxy(trifluoromethyl)boranuide |
Safety and Handling
TSCA : No