Please login to your online account to display your discounted pricing

Potassium triisopropoxyborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC189181000

 View more versions of this product

Catalog No. AC189181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 228.18
Physical Form Solution
Quantity 100mL
Color Colorless to light yellow
Packaging AcroSeal™ Glass Bottle in Tin Can
Assay 1M (exact strength on the certificate of analysis)
Density 0.9140g/mL
Fieser 09,393; 12,414
Chemical Name or Material Potassium triisopropoxyborohydride
Molecular Formula C9H22BKO3
CAS 42278-67-1
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Reacts viol
MDL Number MFCD00044439
Linear Formula KB[OCH(CH3)2]3H
CAS 109-99-9
Name Note 1M solution in THF
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou

Specifications

Chemical Name or Material Potassium triisopropoxyborohydride
Name Note 1M solution in THF
CAS 42278-67-1
CAS 109-99-9
Assay 1M (exact strength on the certificate of analysis)
Molecular Formula C9H22BKO3
Linear Formula KB[OCH(CH3)2]3H
Formula Weight 228.18
MDL Number MFCD00044439
Accessories

Danger

GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Reacts viol

SDS & Safety Documents