missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium triisopropoxyborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

CAS 42278-67-1,109-99-9
Molecular Formula C9H22BKO3
Linear Formula KB[OCH(CH3)2]3H
Formula Weight 228.18
MDL Number MFCD00044439
Chemical Name or Material Potassium triisopropoxyborohydride, 1M solution in THF
Assay 1M (exact strength on the certificate of analysis)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC189181000 SDS Thermo Fisher Scientific
AC189181000
AcroSeal™ Glass Bottle in Tin Can 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Reduction reagent
Description & Specifications

Chemical Identifiers

42278-67-1,109-99-9
KB[OCH(CH3)2]3H
MFCD00044439
1M (exact strength on the certificate of analysis)
C9H22BKO3
228.18
Potassium triisopropoxyborohydride, 1M solution in THF

Specifications

228.18
100mL
AcroSeal™ Glass Bottle in Tin Can
09,393; 12,414
42278-67-1,109-99-9
KB[OCH(CH3)2]3H
1M (exact strength on the certificate of analysis)
Solution
Colorless to light yellow
0.9140g/mL
C9H22BKO3
MFCD00044439
Potassium triisopropoxyborohydride, 1M solution in THF

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Reacts viol

GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou

Danger