missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Trifluoro(trifluoromethyl)borate, TCI America™

Manufacturer:  TCI America P169225G

 View more versions of this product

Catalog No. P169225G



Specifications

Specifications

Potassium Trifluoro(trifluoromethyl)borate
25g
CBF6K
[B-](C(F)(F)F)(F)(F)F.[K+]
175.91
175.91
≥98.0% (T)
White
42298-15-7
UOGBGCVSVMQVBR-UHFFFAOYSA-N
potassium;trifluoro(trifluoromethyl)boranuide
23674658
Crystal-Powder at 20°C

Chemical Identifiers

42298-15-7
175.91
23674658
[B-](C(F)(F)F)(F)(F)F.[K+]
CBF6K
UOGBGCVSVMQVBR-UHFFFAOYSA-N
potassium;trifluoro(trifluoromethyl)boranuide
Safety and Handling

Safety and Handling

TSCA : No

Documents