missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium tricyanomethanide

Manufacturer:  Alfa Aesar™ 2070206

 View more versions of this product

Catalog No. AA2070206



Specifications

Specifications

Potassium tricyanomethanide
5g
C4KN3
MFCD00058850
potassium tricyanomethanide, potassium methanetricarbonitrile, potassium cyanoform, acmc-1ajhe, potassium ion tricyanomethanide, tricyanomethanide potassium salt, potassiomethanetricarbonitrile
C(#N)[C-](C#N)C#N.[K+]
129.163
129.16
White
34171-69-2
KC(CN)3
UN3439
ZKJPYQKGNUBNOA-UHFFFAOYSA-N
potassium;methanetricarbonitrile
10964525
Crystalline

Chemical Identifiers

34171-69-2
129.163
ZKJPYQKGNUBNOA-UHFFFAOYSA-N
10964525
C(#N)[C-](C#N)C#N.[K+]
C4KN3
MFCD00058850
potassium tricyanomethanide, potassium methanetricarbonitrile, potassium cyanoform, acmc-1ajhe, potassium ion tricyanomethanide, tricyanomethanide potassium salt, potassiomethanetricarbonitrile
potassium;methanetricarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents