Please login to your online account to display your discounted pricing

Potassium tri-sec-butylborohydride, 1M solution in tetrahydrofuran, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC205478000

 View more versions of this product

Catalog No. AC205478000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.
Physical Form Solution
Formula Weight 222.26
Quantity 800mL
Color Clear slightly yellow
Packaging Glass Bottle
Density 0.9100g/mL
Merck Index 15, 8564
Fieser 06,490; 07,307
CAS 109-99-9
Molecular Formula C12H28BK
CAS 54575-49-4
MDL Number MFCD00009324
Chemical Name or Material Potassium tri-sec-butylborohydride
Name Note 1M Solution in tetrahydrofuran
Storage Information Product may darken on storage
Linear Formula KB[CH(CH3)C2H5]3H
Health Hazard 1 Danger

Specifications

Chemical Name or Material Potassium tri-sec-butylborohydride
Name Note 1M Solution in tetrahydrofuran
CAS 54575-49-4
CAS 109-99-9
Molecular Formula C12H28BK
Linear Formula KB[CH(CH3)C2H5]3H
Formula Weight 222.26
MDL Number MFCD00009324
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Highly flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.

SDS & Safety Documents