Please login to your online account to display your discounted pricing

Potassium tri-sec-butylborohydride, 1M solution in tetrahydrofuran, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal glass bottle

Manufacturer: thermo fisher scientific  205471000

 View more versions of this product

Catalog No. AC205471000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 222.26
Quantity 100mL
Color Clear slightly yellow
Fieser 06,490; 07,307
Merck Index 15, 8564
Density 0.9100g/mL
Chemical Name or Material Potassium tri-sec-butylborohydride
Name Note 1M Solution in tetrahydrofuran
Molecular Formula C12H28BK
Linear Formula KB[CH(CH3)C2H5]3H
MDL Number MFCD00009324
CAS 54575-49-4
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou
Packaging AcroSeal glass bottle
Health Hazard 2 GHS P Statement Highly flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts viol
Storage Information Product may darken on storage
CAS 109-99-9

Specifications

Chemical Name or Material Potassium tri-sec-butylborohydride
Name Note 1M Solution in tetrahydrofuran
CAS 54575-49-4
CAS 109-99-9
Molecular Formula C12H28BK
Linear Formula KB[CH(CH3)C2H5]3H
Formula Weight 222.26
MDL Number MFCD00009324
Accessories

Danger

GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou

GHS P Statement Highly flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts viol

SDS & Safety Documents