Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium titanium oxide, 98% (metals basis)

Chemical Identifiers

Chemical Name or Material Potassium titanium oxide
CAS 12030-97-6
Formula Weight 174.06
MDL Number MFCD00058803
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1413822

  SDS
alfa aesar™
1413822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1413836

  SDS
alfa aesar™
1413836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 174.06
Assay Percent Notes (metals basis)
Physical Form Powder
TSCA TSCA
Mesh Size −325 Mesh
Assay 98%
Chemical Name or Material Potassium titanium oxide
Melting Point 1615°C
Quantity 100g
CAS 12030-97-6
Odor Odorless
MDL Number MFCD00058803