missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium thiophene-3-trifluoroborate, 97%

Manufacturer:  Alfa Aesar™ H5531303

 View more versions of this product

Catalog No. AAH5531303



Specifications

Specifications

Potassium thiophene-3-trifluoroborate
C4H3BF3KS
amtb173
[B-](C1=CSC=C1)(F)(F)F.[K+]
190.032
190.04
97%
1g
192863-37-9
MFCD03701626
ULOOPHJJEPUNNH-UHFFFAOYSA-N
potassium;trifluoro(thiophen-3-yl)boranuide
23677379
Powder
White

Chemical Identifiers

192863-37-9
190.032
ULOOPHJJEPUNNH-UHFFFAOYSA-N
23677379
[B-](C1=CSC=C1)(F)(F)F.[K+]
C4H3BF3KS
MFCD03701626
amtb173
potassium;trifluoro(thiophen-3-yl)boranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : ED3367500

TSCA : No

Recommended Storage : Ambient temperatures

Documents