Please login to your online account to display your discounted pricing

Potassium thioglycolate, 40-44 wt.% solution in water, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384700010

 View more versions of this product

Catalog No. AC384700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 130.21
Quantity 1L
Color Clear faintly pink
pH 6.8 to 7.3
Assay 40.0 to 44.0%
Density 1.25g/mL
Name Note 40-44 wt.% solution in water
Molecular Formula C2H3KO2S
MDL Number MFCD00067265
Synonym Potassium mercaptoacetate, 40-44 wt.% solution in water
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
Identification potassium: positive, mercaptane: positive
Iron (Fe) 0.5ppm max.
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Potassium thioglycolate
CAS 34452-51-2
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle

Specifications

Chemical Name or Material Potassium thioglycolate
Name Note 40-44 wt.% solution in water
CAS 34452-51-2
CAS 7732-18-5
Assay 40.0 to 44.0%
Molecular Formula C2H3KO2S
Formula Weight 130.21
MDL Number MFCD00067265
Synonym Potassium mercaptoacetate, 40-44 wt.% solution in water
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

SDS & Safety Documents