missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thioglycolate, 40-44 wt.% solution in water, ACROS Organics™

Chemical Identifiers

CAS 34452-51-2
Molecular Formula C2H3KO2S
Molecular Weight (g/mol) 130.202
MDL Number MFCD00067265
InChI Key HYTYHTSMCRDHIM-UHFFFAOYSA-M
Synonym 68-11-1 parent
PubChem CID 23688957
IUPAC Name potassium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC384702500 SDS View Documents Acros Organics
AC384702500
250mL Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC384700010 SDS View Documents Acros Organics
AC384700010
1L Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

34452-51-2
130.202
HYTYHTSMCRDHIM-UHFFFAOYSA-M
23688957
C(C(=O)[O-])S.[K+]
C2H3KO2S
MFCD00067265
68-11-1 parent
potassium;2-sulfanylacetate

Specifications

34452-51-2,7732-18-5
MFCD00067265
68-11-1 parent
C(C(=O)[O-])S.[K+]
130.202
130.21
40.0 to 44.0%
0.5ppm max.
Solubility in water: soluble.
Pink
250mL
C2H3KO2S
1.2500g/mL
HYTYHTSMCRDHIM-UHFFFAOYSA-M
potassium;2-sulfanylacetate
23688957
Solution
potassium: positive, mercaptane: positive
Glass bottle
1.25
6.8 to 7.3
Potassium thioglycolate, 40-44 wt.% solution in water
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning