Please login to your online account to display your discounted pricing

Potassium Thiocyanate, For Iodide, Certified, 4.0% (w/v) ±0.4%, LabChem™

$64.60 - $64.60

Chemical Identifiers

Chemical Name or Material Potassium Thiocyanate
CAS 7732-18-5
CAS Max % 96
CAS 333-20-0
CAS Max % 4
Name Note For Iodide
Purity Grade Certified Grade
Linear Formula KSCN
Formula Weight 97.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC201301

 
lab chem
LC201301
500mL Each for $64.60
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 4.0% ± 0.4% (w/v)
Formula Weight 97.18
Chemical Name or Material Potassium Thiocyanate
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Color Clear, colorless
Quantity 500mL
Name Note For Iodide
Physical Form Liquid
Linear Formula KSCN
CAS 7732-18-5
CAS Max % 96
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 333-20-0
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 4
Solubility Soluble in water
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.