Please login to your online account to display your discounted pricing

Potassium Thiocyanate ACS, Ricca Chemical


Quantity 500g
Packaging Poly wht WM

Manufacturer: ricca chemical company  RIRDCP0730500B1

 View more versions of this product

Catalog No. RDCP0730500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Purity Grade ACS Grade
Physical Form Crystals
Color Colorless or white
CAS Max % 100.0
CAS Min % 98.0
Name Note Thiocyanate
Chemical Name or Material Potassium
Packaging Poly wht WM
Shelf Life 60 months
CAS 333-20-0

Specifications

Chemical Name or Material Potassium
Name Note Thiocyanate
CAS 333-20-0
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Grade
Physical Form Crystals
Accessories

60 months