missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Thiocyanate ACS, Ricca Chemical

$315.94 - $315.94

Chemical Identifiers

CAS 333-20-0
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Chemical Name or Material Potassium, Thiocyanate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RDCP0730500 Ricca Chemical Company
RIRDCP0730500B1
Poly wht WM 500g Each for $315.94
RDCP073010F Ricca Chemical Company
RIRDCP073010F4
Drum, Fiber 10kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

333-20-0
100.0
Potassium, Thiocyanate
98.0
Crystals
ACS Grade

Specifications

333-20-0
100.0
Poly wht WM
500g
ACS Grade
98.0
Crystals
Colorless or white
Potassium, Thiocyanate

Safety and Handling

ShelfLife : 60 months