Please login to your online account to display your discounted pricing

Potassium Thiocyanate, ACS Grade, ≥ 98.5%, LabChem™

$186.94 - $186.94

Chemical Identifiers

Chemical Name or Material Potassium Thiocyanate
CAS 333-20-0
CAS Max % 100
Purity Grade ACS Grade
Linear Formula KSCN
Formula Weight 97.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC201201

 
lab chem
LC201201
500g Each for $186.94
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 97.18
Chemical Name or Material Potassium Thiocyanate
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystalline
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula KSCN
CAS 333-20-0
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Color White
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.