Please login to your online account to display your discounted pricing

Potassium Thiocyanate ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Thiocyanate
CAS 333-20-0
Purity Grade ACS Grade
Molecular Formula KSCN
Formula Weight 97.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7168125

 
avantor performance
716802
POLYSTORMOR 125g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK7168212

 
avantor performance
716806
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 97.18
Packaging POLYSTORMOR
Chemical Name or Material Potassium Thiocyanate
CAS 333-20-0
Molecular Formula KSCN
Quantity 125g