Please login to your online account to display your discounted pricing

Potassium Thiocyanate ACS AR, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  716802

 View more versions of this product

Catalog No. MK7168125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 97.18
Packaging POLYSTORMOR
Chemical Name or Material Potassium Thiocyanate
CAS 333-20-0
Molecular Formula KSCN
Quantity 125g

Specifications

Chemical Name or Material Potassium Thiocyanate
CAS 333-20-0
Purity Grade ACS Grade
Molecular Formula KSCN
Formula Weight 97.18