missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 196582500

 View more versions of this product

Catalog No. AC196582500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Potassium thiocyanate
p.a.
White
Crystalline Powder
99% min. (Argentometry)
KSCN
01,954
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
516872
97.18
Analytical
1.886
170.0°C to 179.0°C
333-20-0
5.3 to 8.7 (5% soln. at 20°C)
250 g
CKNS
MFCD00011413
15, 7807
Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
C(#N)[S-].[K+]
97.18
CHEBI:30951
≥99%
Plastic bottle
1.8860g/mL

Chemical Identifiers

333-20-0
97.18
ZNNZYHKDIALBAK-UHFFFAOYSA-M
516872
potassium;thiocyanate
CKNS
MFCD00011413
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
CHEBI:30951
C(#N)[S-].[K+]
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-370-1

RTECSNumber : XL1925000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only