Please login to your online account to display your discounted pricing

Potassium Thiocyanate, 4% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Thiocyanate
Name Note 4% (w/v)
CAS 7732-18-5
CAS 333-20-0
Purity Grade APHA Standard Methods Reagent
Physical Form Liquid
Assay Percent Range 4.00 ± 0.05% (w/v)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R655000020F

 
ricca chemical company
RIR655000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R65500004A

 
ricca chemical company
RIR65500004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 20L
Purity Grade APHA Standard Methods Reagent
Name Note 4% (w/v)
Chemical Name or Material Potassium Thiocyanate
CAS 7732-18-5
Solvent or Matrix Aqueous Solution
Assay Percent Range 4.00 ± 0.05% (w/v)
CAS 333-20-0
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite