Please login to your online account to display your discounted pricing

Potassium Thiocyanate, 4% (w/v) Aqueous Solution, Ricca Chemical


Quantity 4L
Packaging Poly natural

Manufacturer: ricca chemical company  RIR65500004A

 View more versions of this product

Catalog No. R65500004A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Purity Grade APHA Standard Methods Reagent
Chemical Name or Material Potassium Thiocyanate
CAS 7732-18-5
Assay Percent Range 4.00 ± 0.05% (w/v)
Solvent or Matrix Aqueous Solution
Name Note 4% (w/v)
CAS 333-20-0
Packaging Poly natural
Solubility Solubility in Water: Infinite
Odor Odorless

Specifications

Chemical Name or Material Potassium Thiocyanate
Name Note 4% (w/v)
CAS 7732-18-5
CAS 333-20-0
Purity Grade APHA Standard Methods Reagent
Physical Form Liquid
Assay Percent Range 4.00 ± 0.05% (w/v)