missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Thiocyanate, 10% (w/v) Aqueous Solution, Ricca Chemical

$87.03 - $128.79

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6555-32 View Documents Ricca Chemical Company
RI655532
1L Plastic Each for $128.79
6555-16 View Documents Ricca Chemical Company
RI655516
500mL Plastic Each for $87.03
6555-1 View Documents Ricca Chemical Company
RI65551
4L Plastic N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7732-18-5
dihydrogen oxide, dihydrogen monoxide

Specifications

7732-18-5
10% (w/v) Aqueous Solution
1L
dihydrogen oxide, dihydrogen monoxide
Plastic
Potassium Thiocyanate