missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thioacetate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC221305000

 View more versions of this product

Catalog No. AC221305000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Potassium thioacetate
Crystalline Powder, Crystals or Chunks
98%
CH3COSK
(10% in water) clear
Authentic
White to light brown
500g
10387-40-3
≥97.5% (Argentometry)
C2H3KOS
114.2
Potassium thiolacetate
Glass bottle
≤11 (10% aq. soln.)

Chemical Identifiers

98%
CH3COSK
Potassium thiolacetate
≥97.5% (Argentometry)
C2H3KOS
114.2
Potassium thioacetate