Learn More
Potassium thioacetate, 98%, Thermo Scientific Chemicals
$98.12 - $286.30
Chemical Identifiers
CAS | 10387-40-3 |
---|---|
Molecular Formula | C2H4KOS |
Molecular Weight (g/mol) | 115.211 |
MDL Number | MFCD00083065 |
InChI Key | SDJHDRMYZQFJJO-UHFFFAOYSA-N |
Synonym | potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium |
PubChem CID | 24201352 |
IUPAC Name | ethanethioic S-acid;potassium |
SMILES | CC(=O)S.[K] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0540514
|
Thermo Scientific Chemicals
L0540514 |
25 g |
Each for $98.12
|
|
|||||
AAL0540522
|
Thermo Scientific Chemicals
L0540522 |
100 g |
Each for $286.30
|
|
|||||
Description
Potassium thioacetate is used for palladium mediated coupling with aryl halides and triflates leading to S-arylthioacetates and derivatives. It is also used as a reagent in the conversion of halides to thiols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium thioacetate is used for palladium mediated coupling with aryl halides and triflates leading to S-arylthioacetates and derivatives. It is also used as a reagent in the conversion of halides to thiols.
Solubility
Soluble in water, methanol and some polar solvents. Insoluble in alkanes, ethers and most nonpolar solvents.
Notes
Air sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Chemical Identifiers
10387-40-3 | |
115.211 | |
SDJHDRMYZQFJJO-UHFFFAOYSA-N | |
24201352 | |
CC(=O)S.[K] |
C2H4KOS | |
MFCD00083065 | |
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium | |
ethanethioic S-acid;potassium |
Specifications
10387-40-3 | |
Stench | |
MFCD00083065 | |
UN3335 | |
Air sensitive; Hygroscopic | |
Soluble in water,methanol and some polar solvents. Insoluble in alkanes,ethers and most nonpolar solvents. | |
CC(=O)S.[K] | |
115.211 | |
114.21 | |
Potassium thioacetate |
∽240°C (decomposition) | |
C2H4KOS | |
25 g | |
3595448 | |
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium | |
SDJHDRMYZQFJJO-UHFFFAOYSA-N | |
ethanethioic S-acid;potassium | |
24201352 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
EINECSNumber : 233-848-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only