missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium tetraphenylborate, 97%

Manufacturer:  Alfa Aesar™ H5527614

 View more versions of this product

Catalog No. AAH5527614



Description

Description

Applications
It is an active pharmaceutical intemediate.

Solubility
Low solubility in water. Soluble in organic solvents.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

Potassium tetraphenylborate
C24H20BK
MFCD03093877
MAMCUZQNCMYPCY-UHFFFAOYSA-N
potassium;tetraphenylboranuide
23665558
Powder
White
25g
3244-41-5
KB(C6H5)4
potassium tetrakis phenyl borate
[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[K+]
358.332
358.33
97%
>300°C

Chemical Identifiers

3244-41-5
358.332
MAMCUZQNCMYPCY-UHFFFAOYSA-N
23665558
[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[K+]
C24H20BK
MFCD03093877
potassium tetrakis phenyl borate
potassium;tetraphenylboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents