missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium tetranitropalladate(II), Pd 28.3% min

Manufacturer:  Alfa Aesar™ 1053703

 View more versions of this product

Catalog No. AA1053703



Specifications

Specifications

Potassium tetranitropalladate(II)
K2N4O8Pd-2
dipotassium tetranitropalladiumdiuide, potassium tetranitropalladate ii , pd, palladate 2-,tetrakis nitrito-kn-,potassium 1:2 , sp-4-1
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[K+].[K+].[Pd]
368.637
368.63
Pd 28.3% min.
1g
13844-89-8
MFCD00049788
QZDIEKDUJYMSIK-UHFFFAOYSA-J
dipotassium;palladium;tetranitrite
18763730
Crystalline
310°C (decomposition)

Chemical Identifiers

13844-89-8
368.637
QZDIEKDUJYMSIK-UHFFFAOYSA-J
18763730
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[K+].[K+].[Pd]
K2N4O8Pd-2
MFCD00049788
dipotassium tetranitropalladiumdiuide, potassium tetranitropalladate ii , pd, palladate 2-,tetrakis nitrito-kn-,potassium 1:2 , sp-4-1
dipotassium;palladium;tetranitrite
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents