missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Tetrakis(4-chlorophenyl)borate 98.0+%, TCI America™
$243.87 - $821.21
Chemical Identifiers
CAS | 14680-77-4 |
---|---|
Molecular Formula | C24H16BCl4K |
Molecular Weight (g/mol) | 496.10 |
MDL Number | MFCD00043105 |
InChI Key | VHPYLDPFVZBNQT-UHFFFAOYSA-N |
Synonym | potassium tetrakis 4-chlorophenyl borate, tetrakis 4-chlorophenyl boron potassium, k-tcpb, potassium tetrakis 4-chlorophenyl boranuide, potassium tetrakis 4-chlorophenyl borate, selectophore tm |
PubChem CID | 23679691 |
IUPAC Name | boron(3+) potassium tetrakis(3-chlorobenzen-1-ide) |
SMILES | [B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1 |
Chemical Identifiers
14680-77-4 | |
496.10 | |
VHPYLDPFVZBNQT-UHFFFAOYSA-N | |
23679691 | |
[B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1 |
C24H16BCl4K | |
MFCD00043105 | |
potassium tetrakis 4-chlorophenyl borate, tetrakis 4-chlorophenyl boron potassium, k-tcpb, potassium tetrakis 4-chlorophenyl boranuide, potassium tetrakis 4-chlorophenyl borate, selectophore tm | |
boron(3+) potassium tetrakis(3-chlorobenzen-1-ide) |
Specifications
14680-77-4 | |
1 g | |
C24H16BCl4K | |
potassium tetrakis 4-chlorophenyl borate, tetrakis 4-chlorophenyl boron potassium, k-tcpb, potassium tetrakis 4-chlorophenyl boranuide, potassium tetrakis 4-chlorophenyl borate, selectophore tm | |
[B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1 | |
496.10 | |
496.10 | |
Potassium Tetrakis(4-chlorophenyl)borate [Anion for the neutral carrier type ion electrode] |
White-Yellow | |
Crystal-Powder at 20°C | |
MFCD00043105 | |
VHPYLDPFVZBNQT-UHFFFAOYSA-N | |
boron(3+) potassium tetrakis(3-chlorobenzen-1-ide) | |
23679691 | |
≥98.0% (HPLC,W) |
Safety and Handling
TSCA : No