Learn More
Potassium tetrafluoroborate, 98%, Thermo Scientific Chemicals
$56.66 - $736.36
Chemical Identifiers
CAS | 14075-53-7 |
---|---|
Molecular Formula | BF4K |
Molecular Weight (g/mol) | 125.90 |
MDL Number | MFCD00011395 |
InChI Key | AKEBROIVCDHVSD-UHFFFAOYSA-N |
Synonym | potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride |
PubChem CID | 518872 |
IUPAC Name | potassium tetrafluoroboranuide |
SMILES | [K+].F[B-](F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1560530
|
Thermo Scientific Chemicals
A1560530 |
250 g |
Each for $56.66
|
|
|||||
AAA156050B
|
Thermo Scientific Chemicals
A156050B |
1000 g |
Each for $157.78
|
|
|||||
AAA156050I
|
Thermo Scientific Chemicals
A156050I |
5000 g |
Each for $736.36
|
|
|||||
Description
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
Solubility
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
Chemical Identifiers
14075-53-7 | |
125.90 | |
AKEBROIVCDHVSD-UHFFFAOYSA-N | |
518872 | |
[K+].F[B-](F)(F)F |
BF4K | |
MFCD00011395 | |
potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride | |
potassium tetrafluoroboranuide |
Specifications
(decomposition) | |
14075-53-7 | |
BF4K | |
MFCD00011395 | |
potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride | |
AKEBROIVCDHVSD-UHFFFAOYSA-N | |
potassium tetrafluoroboranuide | |
518872 | |
98% | |
Hygroscopic | |
Potassium tetrafluoroborate |
529°C | |
250 g | |
KBF4 | |
14,7686 | |
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol. | |
[K+].F[B-](F)(F)F | |
125.90 | |
125.91 | |
1.3245 | |
2.5 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
EINECSNumber : 237-928-2
RTECSNumber : ED2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only