Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium tetracyanopalladate(II) hydrate

Chemical Identifiers

Chemical Name or Material Potassium tetracyanopalladate(II) hydrate
CAS 14516-46-2
Formula Weight 288.68 (Anhydrous)
MDL Number MFCD00150431
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1053503

  SDS
alfa aesar™
1053503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 288.68 (Anhydrous)
Color White
TSCA TSCA
Chemical Name or Material Potassium tetracyanopalladate(II) hydrate
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Crystalline
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Quantity 1g
CAS 14516-46-2
MDL Number MFCD00150431
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.