Please login to your online account to display your discounted pricing

Potassium tetrachloropalladate(II), min 32.0% Pd, ACROS Organics™


Quantity 2g
Packaging Glass bottle

Manufacturer: fisher scientific acros  195120020

 View more versions of this product

Catalog No. AC195120020

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥32.0% (Pd)
Physical Form Crystals or Crystalline Powder
Formula Weight 326.41
Quantity 2g
Color Green-brown to dark brown
EINECS Number 233-049-3
TSCA TSCA
RTECS Number TT4100000
Density 2.67g/mL
Iron (Fe) 10ppm max.
CAS 10025-98-6
Packaging Glass bottle
Calcium (Ca) 10ppm max.
Rhodium (Rh) 20ppm max.
Ruthenium (Ru) 20ppm max.
Chemical Name or Material Potassium tetrachloropalladate(II)
Name Note min 32.0% Pd
Molecular Formula Cl4K2Pd
MDL Number MFCD00011373
Linear Formula K2PdCl4
Gold (Au) 20ppm max.
Silicon (Si) 30ppm max.
Copper (Cu) 10ppm max.
Platinum (Pt) 20ppm max.
Magnesium (Mg) 10ppm max.
Silver (Ag) 20ppm max.
Iridium (Ir) 20ppm max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Potassium tetrachloropalladate(II)
Name Note min 32.0% Pd
CAS 10025-98-6
Assay ≥32.0% (Pd)
Molecular Formula Cl4K2Pd
Linear Formula K2PdCl4
Formula Weight 326.41
MDL Number MFCD00011373
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents