missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetrachloropalladate(II), 99.99% (metals basis), Pd 32.2% min., Alfa Aesar™ Premion™

Manufacturer:  Alfa Aesar 1188504

 View more versions of this product

Catalog No. AA1188504



Specifications

Specifications

Potassium tetrachloropalladate(II)
10025-98-6
Cl4K2Pd
Red-brown crystals
LGCKLDWLSVFMGL-UHFFFAOYSA-J
dipotassium;tetrachloropalladium(2-)
61438
Crystalline
Red to Brown
Pd 32.2% min.
99.99% (metals basis)
MFCD00011373
ccris 6625
Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]
326.417
326.42
Odorless
2g

Chemical Identifiers

10025-98-6
326.417
LGCKLDWLSVFMGL-UHFFFAOYSA-J
61438
Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]
Cl4K2Pd
MFCD00011373
ccris 6625
dipotassium;tetrachloropalladium(2-)
Safety and Handling

Safety and Handling

H290-H319-H302: May be corrosive to metals. Causes serious eye irritation. Harmful if swallowed.

EINECSNumber : 233-049-3

Documents