Please login to your online account to display your discounted pricing

Potassium tetrachloropalladate(II), 99.99% (metals basis), Pd 32.2% min., Alfa Aesar™ Premion™

Chemical Identifiers

Chemical Name or Material Potassium tetrachloropalladate(II)
Name Note Pd 32.2% min.
CAS 10025-98-6
Physical Form Crystalline
Assay Percent Range 99.99% (metals basis)
Molecular Formula K2PdCl4
Formula Weight 326.42
MDL Number MFCD00011373
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1188504

  SDS
alfa aesar
1188504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1188509

  SDS
alfa aesar
1188509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 326.42
Quantity 2g
Color Red-brown
Physical Form Crystalline
Chemical Name or Material Potassium tetrachloropalladate(II)
EINECS Number 233-049-3
Odor Odorless
Name Note Pd 32.2% min.
Assay Percent Range 99.99% (metals basis)
Molecular Formula K2PdCl4
Appearance Red-brown crystals
MDL Number MFCD00011373
CAS 10025-98-6
Health Hazard 1 H290-H319-H302: May be corrosive to metals. Causes serious eye irritation. Harmful if swallowed.
Safety and Handling

H290-H319-H302: May be corrosive to metals. Causes serious eye irritation. Harmful if swallowed.