Learn More
Alfa Aesar™ Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min
Manufacturer: Alfa Aesar™ 1215003
Specifications
Potassium tetrachloroaurate(III) hydrate | |
(metals basis) | |
KAuCl4·xH2O | |
UN3260 | |
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion | |
BGVPYGKVUIFYCE-UHFFFAOYSA-J | |
potassium;tetrachlorogold(1-);hydrate | |
91886546 | |
Crystalline | |
Premion™ | |
Yellow to Orange | |
1g |
27988-75-6 | |
AuCl4H2KO | |
MFCD00043088 | |
14,7681 | |
Soluble in water | |
O.Cl[Au-](Cl)(Cl)Cl.[K+] | |
395.88 | |
377.88 (Anhydrous) | |
99.99% | |
Au 49% min. | |
357°C (decomposition) |
Chemical Identifiers
27988-75-6 | |
395.88 | |
BGVPYGKVUIFYCE-UHFFFAOYSA-J | |
91886546 | |
O.Cl[Au-](Cl)(Cl)Cl.[K+] |
AuCl4H2KO | |
MFCD00043088 | |
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion | |
potassium;tetrachlorogold(1-);hydrate |
Safety and Handling
GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA : TSCA