Learn More
Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min, Thermo Scientific Chemicals
$243.68 - $1222.12
Chemical Identifiers
CAS | 27988-75-6 |
---|---|
Molecular Formula | AuCl4K |
Molecular Weight (g/mol) | 377.86 |
MDL Number | MFCD00043088 |
InChI Key | IKHCGJWURGSQRT-UHFFFAOYSA-J |
Synonym | potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion |
PubChem CID | 91886546 |
IUPAC Name | potassium;tetrachlorogold(1-);hydrate |
SMILES | [K+].Cl[Au+3](Cl)(Cl)Cl |
Description
Potassium tetrachloroaurate(III) hydrate is used as a raw material for the preparation of gold(III) dithiolate complexes, which finds application in luminescence studies, photosensitizers and photocatalysts. Further, it is used for gold electroplating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27988-75-6 | |
377.86 | |
IKHCGJWURGSQRT-UHFFFAOYSA-J | |
91886546 | |
[K+].Cl[Au+3](Cl)(Cl)Cl |
AuCl4K | |
MFCD00043088 | |
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion | |
potassium;tetrachlorogold(1-);hydrate |
Specifications
357°C (decomposition) | |
Yellow to Orange | |
1 g | |
KAuCl4·xH2O | |
UN3260 | |
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion | |
IKHCGJWURGSQRT-UHFFFAOYSA-J | |
potassium;tetrachlorogold(1-);hydrate | |
91886546 | |
99.99% | |
Au 49% min. | |
(metals basis) |
27988-75-6 | |
Crystalline | |
AuCl4K | |
MFCD00043088 | |
14,7681 | |
Soluble in water. | |
[K+].Cl[Au+3](Cl)(Cl)Cl | |
377.86 | |
377.88 (Anhydrous) | |
Premion™ | |
Potassium tetrachloroaurate(III) hydrate |
Safety and Handling
GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 237-190-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only